Germanium, AAS standard solution, Specpure®, Ge 1000µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

88067-AE 88067-AP
CAAA88067-AEEA 59.88 CAD
CAAA88067-AE CAAA88067-AP
Germanium, AAS standard solution, Specpure®, Ge 1000µg/ml, Thermo Scientific
Standards Cationic Standards Germanium Standards

Molecular Formula: Ge in 5% HNO3/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085310

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR