Gallium, AAS standard solution, Specpure®, Ga 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88066-AE
88066-AP
CAAA88066-AEEA
72.36
CAD
CAAA88066-AE
CAAA88066-AP
Gallium, AAS standard solution, Specpure®, Ga 1000µg/ml, Thermo Scientific
Standards
Cationic Standards
Gallium Standards
Molecular Formula: Ga in 5% HNO3/tr. HCl
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134045
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...