Gadolinium, AAS standard solution, Specpure®, Gd 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88065-AE
88065-AP
CAAA88065-AEEA
68.57
CAD
CAAA88065-AE
CAAA88065-AP
Gadolinium, AAS standard solution, Specpure®, Gd 1000µg/ml, Thermo Scientific
Standards
Cationic Standards
Gadolinium Standards
Molecular Formula: Gd2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011022
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...