Erbium, AAS standard solution, Specpure®, Er 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88063-AE
88063-AP
CAAA88063-AEEA
64.31
CAD
CAAA88063-AE
CAAA88063-AP
Erbium, AAS standard solution, Specpure®, Er 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Erbium Standards
Molecular Formula: Er2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010987
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...