Cobalt, AAS standard solution, Specpure®, Co 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88060-AE
88060-AP
CAAA88060-AEEA
74.1
CAD
CAAA88060-AE
CAAA88060-AP
Cobalt, AAS standard solution, Specpure®, Co 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Cobalt Standards
Molecular Formula: Co in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010935
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...