Chromium, AAS standard solution, Specpure®, Cr 1000 mg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

88059-AE 88059-AP
CAAA88059-AEEA 67.62 CAD
CAAA88059-AE CAAA88059-AP
Chromium, AAS standard solution, Specpure®, Cr 1000 mg/ml, Thermo Scientific
Standards Cationic Standards Chromium Standards

Molecular Formula: Cr in 5% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00010944

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR