Cesium, AAS Standard Solution, Specpure®, Cs 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

88058-AE 88058-AP
CAAA88058-AEEA 66.2 CAD
CAAA88058-AE CAAA88058-AP
Cesium, AAS Standard Solution, Specpure®, Cs 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Cesium Standards

Molecular Formula: Cs2CO3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134037

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR