Calcium, AAS Standard Solution, Specpure®, Ca 1000 mg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88057-AE
88057-AP
CAAA88057-AEEA
65.89
CAD
CAAA88057-AE
CAAA88057-AP
Calcium, AAS Standard Solution, Specpure®, Ca 1000 mg/ml, Thermo Scientific
Standards
Cationic Standards
Calcium Standards
Molecular Formula: CaCO3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085314
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...