Cadmium, AAS Standard Solution, Specpure®, Cd 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88056-AE
88056-AP
CAAA88056-AEEA
68.89
CAD
CAAA88056-AE
CAAA88056-AP
Cadmium, AAS Standard Solution, Specpure®, Cd 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Cadmium Standards
Molecular Formula: Cd in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010914
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...