Boron, AAS Solution standard, Specpure®, B 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88055-AE
88055-AP
CAAA88055-AEEA
64.46
CAD
CAAA88055-AE
CAAA88055-AP
Boron, AAS Solution standard, Specpure®, B 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Boron Standards
Molecular Formula: H3BO3 in H2O
Physical Form: Liquid
MDL No.: MFCD00134034
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...