Bis(cyclopentadienyl)nickel, dry
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Nickelocene
87899-04
87899-18
CAAA87899-04EA
73
CAD
CAAA87899-04
CAAA87899-18
Bis(cyclopentadienyl)nickel, dry
Bis(cyclopentadienyl)nickel
Sublimes in vacuum
CAS No.: 1271-28-9
Molecular Formula: C10H10Ni
Formula Weight: 188.90
Physical Form: Crystalline, Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Dark green
Melting Point: 171-173°
UN No.: 3391
MDL No.: MFCD00001441
Formula:
C₁₀H₁₀Ni MW: 188.88 g/mol Melting Pt: 171…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001441 CAS Number: 1271-28-9 EINECS: 215-039-0 UN: 1325 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...