Ethylmagnesium bromide 3 M Stabilised with diethyl ether
Supplier: Thermo Scientific Chemicals
|
Danger
|
87291-JT
87291-JU
CAAA87291-JTEA
109.18
CAD
CAAA87291-JT
CAAA87291-JU
Ethylmagnesium bromide 3 M Stabilised with diethyl ether
Ethylmagnesium bromide
Applications: Grignard reagent
CAS No.: 925-90-6
Molecular Formula: C2H5BrMg
Formula Weight: 133.28
Physical Form: Liquid
Density: 1.020
Sensitivity: Air and Moisture Sensitive
Appearance: Pale yellow to brown
Odor: Ether-like
Melting Point: -116.3°
Boiling Point: 34.6°
UN No.: 3399
MDL No.: MFCD00000043
Grignard reagent
Formula:
C₂H₅BrMg MW: 133.27 g/mol Boiling Pt: 34.6 °C Melting Pt: –116.3 °C Density: 1.020 Storage Temperature: Ambient |
MDL Number:
MFCD00000043 CAS Number: 925-90-6 EINECS: 213-127-3 UN: 3399 ADR: 4.3 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...