Fullerene C60 99%, powder

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Fullerene - C60, Buckminster-Fullerene C60, C60 Fullerene, Fullerene-C60, Bucky Ball, Footballen

046311-03 046311-06 046311-MD
AA46311-03EA 492.96 CAD
AA46311-03 AA46311-06 AA46311-MD
Fullerene C60 99%, powder
Fullerene C60

Purity: 99% CAS No.: 99685-96-8 Density: 1.7 Appearance: Black Boiling Point: 500-600° subl. UN No.: 1325 MDL No.: MFCD00282904

Formula: C₆₀
Boiling Pt: 500…600 °C (subl.)
Melting Pt: > 280 °C
Density: 1.7
Storage Temperature: Ambient
MDL Number: MFCD00151408
CAS Number: 99685-96-8
UN: 1325
ADR: 4.1,III
Merck Index: 13,01447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2c9ff03e3743861d242e7e023c84fcfa10a64ac&vid=53371479acfcc1403d111c470b60d1af20934c57&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR