Manganese(II) perchlorate hexahydrate ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
44318-09
44318-18
CAAA44318-09EA
77.89
CAD
CAAA44318-09
CAAA44318-18
CAAA44318-30
Manganese(II) perchlorate hexahydrate ≥99.995% (metals basis)
Manganese (II) perchlorate hexahydrate
Purity: 99.995%
CAS No.: 15364-94-0
Molecular Formula: Mn(ClO4)2.6H2O
Formula Weight: 361.93 (253.84anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
UN No.: 1481
MDL No.: MFCD00150255
Formula:
Cl₂MnO₈·6H₂O Melting Pt: 165 °C. (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00150255 CAS Number: 15364-94-0 EINECS: 237-390-9 UN: 1481 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...