Ethylene glycol dimethacrylate (ethylene dimethacrylate) 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) 100 ppm

44151-18 44151-30 44151-A1 44151-A6
CAAA44151-18EA 45.19 CAD
CAAA44151-18 CAAA44151-30 CAAA44151-A1 CAAA44151-A6
Ethylene glycol dimethacrylate (ethylene dimethacrylate) 98% stabilized
Ethylene glycol dimethacrylate (Ethylene dimethacrylate)

Applications: Crosslinking monomer


Purity: 98% CAS No.: 97-90-5 Molecular Formula: C10H14O4 Formula Weight: 198.21 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.051 Refractive Index: 1.4540 Appearance: Colorless to pale yellow Boiling Point: 83-85°/1mm Beilstein Reference: 1776663 MDL No.: MFCD00008590


Crosslinking monomer

Formula: C₁₀H₁₄O₄
Boiling Pt: 83…85 °C (1 mmHg)
Density: 1.051
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008590
CAS Number: 97-90-5
EINECS: 202-617-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR