MOPSO (3-(4-morpholino)-2-hydroxypropanesulfonic acid) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
44135-18
44135-30
44135-A1
CAAA44135-09EA
42.94
CAD
CAAA44135-09
CAAA44135-18
CAAA44135-30
CAAA44135-A1
MOPSO (3-(4-morpholino)-2-hydroxypropanesulfonic acid) 98%
MOPSO (3-(4-Morpholino)-2-hydroxypropanesulphonic acid)
Biological buffers, pH range 6.2-7.6
Purity: 98%
CAS No.: 68399-77-9
Molecular Formula: C7H15NO5S
Formula Weight: 225.27
Physical Form: Powder
Melting Point: 276-279°
MDL No.: MFCD00009629
Biological buffers, pH range 6.2-7.6
Formula:
C₇H₁₅NO₅S Melting Pt: 276…279 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009629 CAS Number: 68399-77-9 EINECS: 269-989-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...