(2-Hydroxyethyl)hydrazine
Supplier: Thermo Scientific Chemicals
|
Danger
|
43793-09
43793-18
43793-30
CAAA43793-09EA
58.46
CAD
CAAA43793-09
CAAA43793-18
CAAA43793-30
(2-Hydroxyethyl)hydrazine
(2-Hydroxyethyl)hydrazine
CAS No.: 109-84-2
Molecular Formula: HOCH2CH2NHNH2
Formula Weight: 76.10
Flash Point: 73°(163°F)
Physical Form: Liquid
Density: 1.121
Refractive Index: 1.4930
Melting Point: -70°
Boiling Point: 218-220°
UN No.: 2810
MDL No.: MFCD00007623
Formula:
C₂H₈N₂O Boiling Pt: 218…220 °C Melting Pt: –70 °C Density: 1.121 Flash Pt: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007623 CAS Number: 109-84-2 EINECS: 203-711-6 UN: 2810 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...