Hydroxypropyl cellulose, 75 - 150 mPa.s (5 % in H₂O)
Supplier: Thermo Scientific Chemicals
43400-14
43400-22
43400-36
43400-A3
CAAA43400-14EA
64.15
CAD
CAAA43400-14
CAAA43400-22
CAAA43400-36
CAAA43400-A3
Hydroxypropyl cellulose, 75 - 150 mPa.s (5 % in H₂O)
Hydroxypropyl cellulose
Uses: Thickener, binder, and emulsifier
Average MW 100,000; viscosity (5 wt% aqueous solution, 25°) 75-150 cps; As ³3ppm, Pb ³10ppm
Soluble in polar organic solvents, in water at room temperature
CAS No.: 9004-64-2
Physical Form: Powder
Odor: Odorless
pH: 7
Solubility: Soluble in polar organic solvents, in water at room temperature
Merck Reference: 14,4841
MDL No.: MFCD00132688
Thickener, binder, and emulsifier
Storage Temperature:
Ambient |
MDL Number:
MFCD00132688 CAS Number: 9004-64-2 Merck Index: 14,4841 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...