Trimethylamine 45% (w/w) in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
43282-AP
43282-K4
CAAA43282-APEA
157.05
CAD
CAAA43282-AP
CAAA43282-K4
Trimethylamine 45% (w/w) in aqueous solution
Trimethylamine
CAS No.: 75-50-3
Molecular Formula: (CH3)3N
Formula Weight: 59.11
Flash Point: -20°(-4°F)
Physical Form: Liquid
Appearance: Colorless
Odor: Unpleasant
Melting Point: -2°
Boiling Point: 30°
Beilstein Reference: 956566
Merck Reference: 14,9710
UN No.: 1297
MDL No.: MFCD00008327
Formula:
C₃H₉N Boiling Pt: 30 °C Melting Pt: –2 °C Density: 0.88 g/cm³ (20 °C) Flash Pt: –20 °C (–4 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008327 CAS Number: 75-50-3 EINECS: 200-875-0 UN: 1297 ADR: 3,I Merck Index: 14,9710 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...