Methacrolein ≥96% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with hydroquinone (HQ)
43124-AA
43124-AC
CAAA43124-AAEA
107.76
CAD
CAAA43124-AA
CAAA43124-AC
Methacrolein ≥96% stabilized
Methacrolein
Purity: 96%
CAS No.: 78-85-3
Molecular Formula: C4H6O
Formula Weight: 70.09
Flash Point: -15°(5°F)
Physical Form: Liquid
Density: 0.847
Refractive Index: 1.4150
Sensitivity: Light Sensitive
Appearance: Pale yellow
Odor: Pungent
Melting Point: -81°
Boiling Point: 69-70°
UN No.: 2396
MDL No.: MFCD00006974
Formula:
C₄H₆O Boiling Pt: 69…70 °C Melting Pt: –81 °C Density: 0.847 Flash Pt: –15 °C (5 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006974 CAS Number: 78-85-3 EINECS: 201-150-1 UN: 2396 ADR: 3,II |
Specification Test Results
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...