Divinylbenzene (mixture of isomers) 55% stabilised

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DVB, Divinylbenzene

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC) 1000 ppm

42805-18 42805-30 42805-A1
CAAA42805-18EA 49.14 CAD
CAAA42805-18 CAAA42805-30 CAAA42805-A1
Divinylbenzene (mixture of isomers) 55% stabilised
Divinylbenzene (mixture of isomers)

Applications: Crosslinking monomer


Purity: 55% CAS No.: 1321-74-0 Molecular Formula: C6H4(CH=CH2)2 Formula Weight: 130.19 Flash Point: 62°(143°F) Physical Form: Liquid Density: 0.912 Refractive Index: 1.5740 MDL No.: MFCD01778656


Crosslinking monomer

Formula: C₁₀H₁₀
Density: 0.912
Flash Pt: 62 °C (143 °F)
Storage Temperature: Ambient
MDL Number: MFCD01778656
CAS Number: 1321-74-0
EINECS: 215-325-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR