Divinylbenzene (mixture of isomers) ≥80% stabilised

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DVB, Divinylbenzene

Stabilization: 1000 ppm Stabilised with 4-tert-butylpyrocatechol (TBC)

42804-14 42804-22
CAAA42804-14EA 46.77 CAD
CAAA42804-14 CAAA42804-22 CAAA42804-36
Divinylbenzene (mixture of isomers) ≥80% stabilised
Divinylbenzene (mixture of isomers)

Purity: 80% CAS No.: 1321-74-0 Molecular Formula: C6H4(CH=CH2)2 Formula Weight: 130.19 Storage Temperature: 2°C to 8°C Flash Point: 69°(156°F) Physical Form: Liquid Density: 0.914 Refractive Index: 1.5740 Sensitivity: Light Sensitive Appearance: Colorless to pale yellow MDL No.: MFCD01778656

Formula: C₁₀H₁₀
MW: 130.19 g/mol
Density: 0.914
Flash Pt: 69 °C (156 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD01778656
CAS Number: 1321-74-0
EINECS: 215-325-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR