Alizarin red S (sodium salt) 1% (w/v) in aqueous solution
Supplier: Thermo Scientific Chemicals
Synonyms:
Alizarin sulfonic acid disodium salt, Sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulphonate, C.I. 58005, Sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate, Alizarin sulphonic acid disodium salt, Alizarin Red S sodium
42746-AE
42746-AP
CAAA42746-AEEA
45.66
CAD
CAAA42746-AE
CAAA42746-AP
Alizarin red S (sodium salt) 1% (w/v) in aqueous solution
Alizarin Red S (sodium salt)
Transition interval: pH 4.6 (yellow) to pH 6.0 (red)
CAS No.: 130-22-3
Molecular Formula: C14H7O7SNa
Formula Weight: 342.27
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00013049
Formula:
C₁₄H₇NaO₇S MW: 342.26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013049 CAS Number: 130-22-3 EINECS: 204-981-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...