3-Hydroxy-3-methyl butyric acid ≥96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Hydroxyisovaleric acid, 3-Hydroxy-3-methylbutanoic acid
42722-09
42722-18
42722-30
CAAAA42722-09EA
74.89
CAD
CAAAA42722-09
CAAA42722-18
CAAA42722-30
CAAA42722-A1
3-Hydroxy-3-methyl butyric acid ≥96%
3-Hydroxy-3-methyl butyric acid
Soluble in water
Purity: 96%
CAS No.: 625-08-1
Molecular Formula: C5H10O3
Formula Weight: 118.13
Flash Point: 113°(235°F)
Physical Form: Liquid
Density: 1.1
Refractive Index: 1.4408
Appearance: Pale yellow
Boiling Point: 128°/7mm
Solubility: Soluble in water
Beilstein Reference: 1743952
MDL No.: MFCD00059081
Formula:
C₅H₁₀O₃ Boiling Pt: 128 °C (7 mmHg) Density: 1,1 Flash Pt: 113 °C (235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00059081 CAS Number: 625-08-1 |
Specification Test Results
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...