Abietic acid 90+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
42582-03
42582-MD
CAAA42582-03EA
491.38
CAD
CAAA42582-03
CAAA42582-MD
Abietic acid 90+%
Abietic acid
Purity: 90+%
CAS No.: 514-10-3
Molecular Formula: C20H30O2
Formula Weight: 302.45
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline
Sensitivity: Air Sensitive
Appearance: Pale yellow
Odor: Characteristic
Melting Point: 148-152°
Beilstein Reference: 2221451
Merck Reference: 14,7
MDL No.: MFCD03423567
Formula:
C₂₀H₃₀O₂ Melting Pt: 148…152 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03423567 CAS Number: 514-10-3 EINECS: 208-178-3 Merck Index: 14,7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...