Palladium (II) sulfate dihydrate ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
42529-03
42529-06
CAAA42529-03EA
271.76
CAD
CAAA42529-03
CAAA42529-06
Palladium (II) sulfate dihydrate ≥99% (metals basis)
Palladium (II) sulphate dihydrate
Soluble in cold water; decomposes in hot water
Purity: 99%
CAS No.: 13444-98-9
Molecular Formula: O4PdS.2H2O
Formula Weight: 238.51 (202.48anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Red to brown
Melting Point: dec.
Solubility: Soluble in cold water; decomposes in hot water
UN No.: 3260
MDL No.: MFCD00011173
Formula:
O₄PdS·2H₂O Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00011173 CAS Number: 13444-98-9 EINECS: 236-957-8 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...