Ethylene glycol-[D₆] (98% D)
Supplier: Thermo Scientific Chemicals
|
Warning
|
42288-03
42288-06
CAAA42288-03EA
516.66
CAD
CAAA42288-03
CAAA42288-06
Ethylene glycol-[D₆] (98% D)
Ethylene glycol-[D6]
Purity: 98%
CAS No.: 15054-86-1
Molecular Formula: DOCD2CD2OD
Formula Weight: 68.02
Flash Point: 111°(232°F)
Physical Form: Liquid
Density: 1.189
Odor: Odorless
Boiling Point: 86-87°/8mm
MDL No.: MFCD00078666
Formula:
C₂D₆O₂ Boiling Pt: 86…87 °C (8 mmHg) Density: 1,189 Flash Pt: 111 °C (232 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00078666 CAS Number: 15054-86-1 EINECS: 239-122-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...