1,2-Dichloroethane-[D₄] (99% D)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,2-Dichloroethane-[²H₄]
42281-03
42281-06
42281-46
CAAA42281-03EA
267.02
CAD
CAAA42281-03
CAAA42281-06
CAAA42281-46
1,2-Dichloroethane-[D₄] (99% D)
1,2-Dichloroethane-[D4]
1g size packaged in prescored ampoules
Purity: 99%
CAS No.: 17060-07-0
Molecular Formula: Cl(CD2)2Cl
Formula Weight: 102.99
Flash Point: 15°(60°F)
Physical Form: Liquid
Density: 1.307
Refractive Index: 1.4430
Melting Point: -35°
Boiling Point: 83°
UN No.: 1184
MDL No.: MFCD00037531
Formula:
C₂D₄Cl₂ Boiling Pt: 83 °C Melting Pt: –35 °C Density: 1.307 Flash Pt: 15 °C (60 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00037531 CAS Number: 17060-07-0 UN: 1184 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...