1,2-Dibromoethane-[D₄] (99% D)

Supplier: Thermo Scientific Chemicals
Danger

42279-06 42279-14
CAAA42279-06EA 211.72 CAD
CAAA42279-06 CAAA42279-14
1,2-Dibromoethane-[D₄] (99% D)
1,2-Dibromoethane-[D4]

Purity: 99% CAS No.: 22581-63-1 Molecular Formula: BrCD2CD2Br Formula Weight: 191.89 Physical Form: Liquid Density: 2.20 Refractive Index: 1.5360 Odor: Slightly sweetish Melting Point: 9° Boiling Point: 132° UN No.: 1605 MDL No.: MFCD00037499

Formula: BrH₂CCH₂Br
Boiling Pt: 132 °C
Melting Pt: 9 °C
Density: 2.20
Storage Temperature: Ambient
MDL Number: MFCD00037499
CAS Number: 22581-63-1
EINECS: 245-102-8
UN: 1605
ADR: 6.1,I
Merck Index: 13,03830

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR