Tungsten, Plasma Standard Solution, Specpure®, W 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
42249-AD
42249-AE
42249-AP
CAAA42249-ADEA
145.99
CAD
CAAA42249-AD
CAAA42249-AE
CAAA42249-AP
Tungsten, Plasma Standard Solution, Specpure®, W 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Tungsten Standards
Molecular Formula: (NH4)2WO4 in H2O
Physical Form: Liquid
MDL No.: MFCD00011461
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...