Tungsten, Plasma Standard Solution, Specpure®, W 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
42248-AD
42248-AE
42248-AP
CAAA42248-ADEA
106.81
CAD
CAAA42248-AD
CAAA42248-AE
CAAA42248-AP
Tungsten, Plasma Standard Solution, Specpure®, W 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Tungsten Standards
Molecular Formula: (NH4)2WO4 in H2O
Physical Form: Liquid
MDL No.: MFCD00011461
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...