Germanium, plasma standard solution, Specpure®, Ge 1000µg/ml, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
42242-AP 42242-AE 42242-AE 42242-AP 42242-AD 42242-AD
CAAA42242-APEA 263.86 CAD
CAAA42242-AP AA42242-AP CAAA42242-AD CAAA42242-AE AA42242-AE AA42242-AD
Germanium, plasma standard solution, Specpure®, Ge 1000µg/ml, Thermo Scientific
Standards Cationic Standards Germanium Standards
Molecular Formula: (NH4)2GeF6 in H2O/trace F-
Physical Form: Liquid
MDL No.: MFCD00085310
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR