Osmium, AAS standard solution, Specpure®, Os 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
42239-AP 42239-AE 42239-AE 42239-AP
CAAA42239-AEEA 172.22 CAD
CAAA42239-AE AA42239-AE AA42239-AP CAAA42239-AP
Osmium, AAS standard solution, Specpure®, Os 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Osmium Standards
Molecular Formula: (NH4)2OsCl6 in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011147
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR