Polyethylene oxide, M.W. 100000

Supplier: Thermo Scientific Chemicals

Synonyms: PEO

42236-14 42236-22 42236-36
CAAA42236-14EA 47.56 CAD
CAAA42236-14 CAAA42236-22 CAAA42236-36
Polyethylene oxide, M.W. 100000
Polyethylene oxide

Polyethylene oxide is used in conjunction with carbon black to form a conductive composite. It serves as thermoplastic material that is readily calendared, extruded, injection molded or cast, to provide even greater formulation and processing flexibility. It also provides binding, thickening, lubricity, water retention, and film formation benefits to deliver excellent performance in various applications viz. pharmaceuticals, mining, paper and cleaning products.


Tg=-67° High viscosity, combustible. Keep away from heat, sparks and open flame. Incompatible with strong acids, strong bases and strong oxidizing agents.
Soluble in toluene, THF, hot water, DMF, CHCl3

Formula: (-CH₂CH₂O-)n
Melting Pt: 65 °C
Density: 1.4539
Storage Temperature: Ambient
MDL Number: MFCD00147970
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR