Bis(triphenylphosphine)nickel(II) bromide 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nickel(II) bromide bis(triphenylphosphine), Dibromobis(triphenylphosphine)nickel(II)

42192-03 42192-06
CAAA42192-03EA 46.93 CAD
CAAA42192-03 CAAA42192-06
Bis(triphenylphosphine)nickel(II) bromide 99%
Bis(triphenylphosphine)nickel (II) bromide

Applications: polymerization catalyst


Purity: 99% CAS No.: 14126-37-5 Molecular Formula: Br2Ni[P(C6H5)3]2 Formula Weight: 743.12 Physical Form: Powder Odor: Odorless MDL No.: MFCD00015865


As a polymerization catalyst

Formula: C₃₆H₃₀Br₂NiP₂
MW: 743.08
Melting Pt: 219…223 °C
Storage Temperature: Ambient
MDL Number: MFCD00015865
CAS Number: 14126-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR