Isobutyl methacrylate ≥99.5% stabilised

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methacrylic acid Isobutyl ester

Stabilization: 10 ppm Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)

41948-AK 41948-K2 41948-K7
CAAA41948-AKEA 53.4 CAD
CAAA41948-AK CAAA41948-K2 CAAA41948-K7
Isobutyl methacrylate ≥99.5% stabilised
Isobutyl methacrylate

Purity: 99.5+% CAS No.: 97-86-9 Molecular Formula: H2C=C(CH3)CO2CH2CH(CH3)2 Formula Weight: 142.20 Flash Point: 46°(115°F) Physical Form: Liquid Density: 0.889 Boiling Point: 155° Beilstein Reference: 1747595 UN No.: 2283 MDL No.: MFCD00008931

Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 155 °C
Density: 0.889
Flash Pt: 46 °C (115 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008931
CAS Number: 97-86-9
EINECS: 202-613-0
UN: 2283
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR