Antimony, plasma standard solution, Specpure®, Sb 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

41682-AD 41682-AE 41682-AP
CAAA41682-ADEA 82.63 CAD
CAAA41682-AD CAAA41682-AE CAAA41682-AP
Antimony, plasma standard solution, Specpure®, Sb 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Antimony Standards

Molecular Formula: Sb in H2O/tartaric acid/tr. HNO3
Physical Form: Liquid
MDL No.: MFCD00134030

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR