trans-Bis(triphenylphosphine)palladium(II) chloride ≥99.95% (metals basis) Pd ≥ 14.7%, Premion®

Supplier: Thermo Scientific Chemicals

Synonyms: trans-Dichlorobis(triphenylphosphine)palladium(II)

41245-03 41245-06 41245-14
CAAA41245-03EA 92.75 CAD
CAAA41245-03 CAAA41245-06 CAAA41245-14
trans-Bis(triphenylphosphine)palladium(II) chloride ≥99.95% (metals basis) Pd ≥ 14.7%, Premion®
trans-Bis(triphenylphosphine)palladium (II) chloride

Purity: 99.95% CAS No.: 13965-03-2 Molecular Formula: PdCl2[P(C6H5)3]2 Formula Weight: 701.91 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Yellow Odor: Odorless Melting Point: ca 310° dec. MDL No.: MFCD00009593

Formula: C₃₆H₃₀Cl₂P₂Pd
MW: 701.91 g/mol
Melting Pt: ∼ 310 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00009593
CAS Number: 13965-03-2
EINECS: 237-744-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR