(Triphenylphosphine)gold(I) chloride ≥99.99% (metals basis) Au ≥ 39.3%, Premion®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Chlorotriphenylphosphine gold(I), Chloro(triphenylphosphine)gold(I), Triphenylphosphinechlorogold

41007-02 41007-03 41007-06
CAAA41007-02EA 224.36 CAD
CAAA41007-02 CAAA41007-03 CAAA41007-06
(Triphenylphosphine)gold(I) chloride ≥99.99% (metals basis) Au ≥ 39.3%, Premion®
(Triphenylphosphine)gold (I) chloride

Purity: 99.99% CAS No.: 14243-64-2 Molecular Formula: AuCl[P(C6H5)3] Formula Weight: 494.71 Physical Form: Powder Odor: Odorless MDL No.: MFCD00009588

Formula: C₁₈H₁₅AuClP
MW: 494.71
Storage Temperature: Ambient
MDL Number: MFCD00009588
CAS Number: 14243-64-2
EINECS: 238-117-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR