Scandium (III) trifluoromethanesulfonate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
40566-MD
40566-03
40566-06
CAAAA40566-MDEA
56.25
CAD
CAAAA40566-MD
CAAA40566-03
CAAA40566-06
Scandium (III) trifluoromethanesulfonate 98%
Scandium (III) trifluoromethanesulphonate
Fieser: 18,317 19,300 20,335 21,387
Purity: 98%
CAS No.: 144026-79-9
Molecular Formula: Sc(OSO2CF3)3
Formula Weight: 492.16
Physical Form: Powder
Sensitivity: Hygroscopic
MDL No.: MFCD00798539
Formula:
C₃F₉O₉S₃Sc Storage Temperature: Ambient |
MDL Number:
MFCD00798539 CAS Number: 144026-79-9 |
Specification Test Results
Form | Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...