Bis(cyclopentadienyl)tungsten dichloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
40521-03
40521-MD
CAAA40521-03EA
406.06
CAD
CAAA40521-03
CAAA40521-MD
Bis(cyclopentadienyl)tungsten dichloride 99%
Bis(cyclopentadienyl)tungsten dichloride
Purity: 99%
CAS No.: 12184-26-8
Molecular Formula: C10H10Cl2W
Formula Weight: 384.95
Physical Form: Powder, Packaged under argon
Sensitivity: Air and Moisture Sensitive
UN No.: 3261
MDL No.: MFCD00269852
Formula:
C₁₀H₁₀Cl₂W Melting Pt: >350 °C Storage Temperature: Ambient |
MDL Number:
MFCD00269852 CAS Number: 12184-26-8 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...