Ammonium tetrachloroaurate hydrate ≥99.99% (metals basis), Premion®

Supplier: Thermo Scientific Chemicals
Danger

40434-03 40434-06
CAAA40434-03EA 271.76 CAD
CAAA40434-03 CAAA40434-06
Ammonium tetrachloroaurate hydrate ≥99.99% (metals basis), Premion®
Ammonium tetrachloroaurate hydrate

Soluble in water and alcohol


Purity: 99.99% CAS No.: 13874-04-9 Molecular Formula: (NH4)AuCl4.xH2O Formula Weight: 356.82(anhy) Physical Form: Crystalline powder Appearance: Yellow-orange Odor: Odorless Solubility: Soluble in water and alcohol UN No.: 3260 MDL No.: MFCD00011503

Formula: H₄AuCl₄N·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00011503
CAS Number: 13874-04-9
EINECS: 250-476-0
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR