Bismuth subsalicylate ≥96%

Supplier: Thermo Scientific Chemicals
Warning

40395-30 40395-A1
CAAA40395-30EA 178.54 CAD
CAAA40395-30 CAAA40395-A1
Bismuth subsalicylate ≥96%
Bismuth subsalicylate

Applications: To impart pearly surface to cellulose-base, polystyrene, and phenolformaldehyde resins. Typical tap bulk density 0.43g/cm³


  • Practically insoluble in water, alcohol


Purity: 96% CAS No.: 14882-18-9 Molecular Formula: C7H5BiO4 (approximate) Formula Weight: 362.09 Physical Form: Powder, typically 2 micron max Odor: Odorless Merck Reference: 14,1285 MDL No.: MFCD00085368


To impart pearly surface to cellulose-base, polystyrene, and phenolformaldehyde resins. Typical tap bulk density 0.43g/cm3

Formula: C₇H₅BiO₄
Storage Temperature: Ambient
MDL Number: MFCD00085368
CAS Number: 14882-18-9
EINECS: 238-953-1
Merck Index: 14,1285

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR