Bis(cyclopentadienyl)titanium dichloride 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Titanocenedichloride, Bis(cyclopentadienyl)titanium(IV) dichloride, Titanocene dichloride

40386-04 40386-09 40386-18
CAAA40386-04EA 39.5 CAD
CAAA40386-04 CAAA40386-09 CAAA40386-18
Bis(cyclopentadienyl)titanium dichloride 99+%
Bis(cyclopentadienyl)titanium dichloride

Fieser: 6,48 13,102 14,120 15,120 16,116 18,364 19,348 20,384 21,433


Purity: 99+% CAS No.: 1271-19-8 Molecular Formula: C10H10Cl2Ti Formula Weight: 249.00 Physical Form: Crystalline, Packaged under argon Density: 1.60 Appearance: Red Odor: Odorless Melting Point: ca 285° dec. Merck Reference: 14,9482 MDL No.: MFCD00003723

Formula: C₁₀H₁₀Cl₂Ti
MW: 248.97 g/mol
Melting Pt: ∼ 285 °C (dec.)
Density: 1.60
Storage Temperature: Ambient
MDL Number: MFCD00003723
CAS Number: 1271-19-8
EINECS: 215-035-9
UN: 3261
ADR: 8 II
Merck Index: 14,9482

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR