Yttrium (III) trifluoromethanesulfonate hydrate ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
40315-04
40315-09
CAAA40315-04EA
50.24
CAD
CAAA40315-04
CAAA40315-09
CAAA40315-18
Yttrium (III) trifluoromethanesulfonate hydrate ≥98%
Yttrium (III) trifluoromethanesulphonate hydrate
CAS No.: 34629-25-9
Molecular Formula: Y(OSO2CF3)3.xH2O
Formula Weight: 536.10(anhy)
Physical Form: Powder
MDL No.: MFCD00209623
Formula:
C₃F₉O₉S₃Y·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00209623 CAS Number: 34629-25-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...