Thulium (III) trifluoromethanesulfonate ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
40313-04
40313-09
CAAA40313-04EA
169.06
CAD
CAAA40313-04
CAAA40313-09
Thulium (III) trifluoromethanesulfonate ≥98%
Thulium (III) trifluoromethanesulphonate
Purity: 98%
CAS No.: 141478-68-4
Molecular Formula: C3F9O9S3Tm
Formula Weight: 616.13
Physical Form: Powder
Formula:
C₃F₉O₉S₃Tm·H₂O MW: 616.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00209617 CAS Number: 141478-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...