Terbium (III) trifluoromethanesulfonate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
40312-04
40312-09
CAAA40312-04EA
130.51
CAD
CAAA40312-04
CAAA40312-09
Terbium (III) trifluoromethanesulfonate 98%
Terbium (III) trifluoromethanesulphonate
Purity: 98%
CAS No.: 148980-31-8
Molecular Formula: Tb(OSO2CF3)3
Formula Weight: 606.12
Physical Form: Powder
Formula:
C₃F₉O₉S₃Tb MW: 606.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00192461 CAS Number: 148980-31-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...