Calcium trifluoromethanesulfonate ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
40134-09
40134-18
40134-22
CAAA40134-09EA
155.79
CAD
CAAA40134-09
CAAA40134-18
CAAA40134-22
Calcium trifluoromethanesulfonate ≥99%
Calcium trifluoromethanesulphonate
Purity: 99% min
CAS No.: 55120-75-7
Molecular Formula: C2CaF6O6S2
Formula Weight: 338.21
Physical Form: Powder
Appearance: White
Melting Point: dec.
MDL No.: MFCD00143911
Formula:
C₂CaF₆O₆S₂ Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00143911 CAS Number: 55120-75-7 EINECS: 232-188-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...