Indium(III) trifluoromethanesulfonate ≥99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
40131-09
40131-18
CAAA40131-09EA
240.16
CAD
CAAA40131-09
CAAA40131-18
Indium(III) trifluoromethanesulfonate ≥99%
Indium (III) trifluoromethanesulphonate
Purity: 99% min
CAS No.: 128008-30-0
Molecular Formula: In(CF3SO3)3
Formula Weight: 562.02
Physical Form: Powder
Sensitivity: Hygroscopic
UN No.: 3261
MDL No.: MFCD00144478
Formula:
C₃F₉InO₉S₃ Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00144478 CAS Number: 128008-30-0 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...