Nickel(II) carbonate (basic) hydrate Ni ≥ 40% , ∼99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

40106-30 40106-A1
CAAA40106-30EA 65.41 CAD
CAAA40106-30 CAAA40106-A1
Nickel(II) carbonate (basic) hydrate Ni ≥ 40% , ∼99.5% (metals basis)
Nickel(II) carbonate (basic) hydrate

Applications: Nickel plating pH adjuster, catalysts, ceramic colors and glazes


Purity: 99.5% CAS No.: 155775-31-8 Molecular Formula: CNiO3.2H2NiO2.xH2O Formula Weight: 304.18(anhy) Physical Form: Powder Appearance: Green Odor: Odorless UN No.: 3077 MDL No.: MFCD00016262


Nickel plating pH adjuster, catalysts, ceramic colors and glazes

Formula: CH₄Ni₃O₇·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00016262
CAS Number: 39430-27-8
EINECS: 240-408-8
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR